Anti-ARHGAP5 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP34759_P050
AVIVARP34759_P050EA 401 EUR
AVIVARP34759_P050
Anti-ARHGAP5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARHGAP5 (Rho GTPase activating protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen