Anti-TRIM41 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP34763_P050
AVIVARP34763_P050EA 401 EUR
AVIVARP34763_P050
Anti-TRIM41 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRIM41 (tripartite motif containing 41)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen