Anti-RORA Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP34767_P050
AVIVARP34767_P050EA 401 EUR
AVIVARP34767_P050
Anti-RORA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RORA (RAR-related orphan receptor A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen