Anti-WDR45B Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP34827_P050
AVIVARP34827_P050EA 401 EUR
AVIVARP34827_P050
Anti-WDR45B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: WDR45B (WD repeat domain 45B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen