Anti-PHF11 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP34848_P050
AVIVARP34848_P050EA 401 EUR
AVIVARP34848_P050
Anti-PHF11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PHF11 (PHD finger protein 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen