Anti-CLIC5 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP35263_P050 ARP35263_T100 ARP35263_25UL
AVIVARP35263_P050EA 401 EUR
AVIVARP35263_P050 AVIVARP35263_T100 AVIVARP35263_25UL
Anti-CLIC5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLIC5 (chloride intracellular channel 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR