Anti-KCNK12 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP35349_P050
AVIVARP35349_P050EA 401 EUR
AVIVARP35349_P050
Anti-KCNK12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KCNK12 (potassium channel, subfamily K, member 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen