Anti-VSX1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP35883_P050
AVIVARP35883_P050EA 401 EUR
AVIVARP35883_P050
Anti-VSX1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VSX1 (visual system homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen