Anti-L3MBTL1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP35895_P050
AVIVARP35895_P050EA 401 EUR
AVIVARP35895_P050
Anti-L3MBTL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: L3MBTL1 (l(3)mbt-like 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen