Anti-ZNF581 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP35921_P050
AVIVARP35921_P050EA 401 EUR
AVIVARP35921_P050
Anti-ZNF581 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF581 (zinc finger protein 581)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR