Anti-ZNF571 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP35923_P050
AVIVARP35923_P050EA 401 EUR
AVIVARP35923_P050
Anti-ZNF571 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF571 (zinc finger protein 571)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR