Anti-DMRTC2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP36091_P050
AVIVARP36091_P050EA 401 EUR
AVIVARP36091_P050
Anti-DMRTC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DMRTC2 (DMRT-like family C2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human