Anti-ZNF660 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP36242_T100
AVIVARP36242_T100EA 349 EUR
AVIVARP36242_T100
Anti-ZNF660 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF660 (zinc finger protein 660)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Dog