Anti-DDX54 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP36498_P050
AVIVARP36498_P050EA 401 EUR
AVIVARP36498_P050
Anti-DDX54 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDX54 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 54)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR