Anti-ANXA2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP36588_T100
AVIVARP36588_T100EA 349 EUR
AVIVARP36588_T100
Anti-ANXA2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ANXA2 (annexin A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen