Anti-EGR1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP36840_P050
AVIVARP36840_P050EA 401 EUR
AVIVARP36840_P050
Anti-EGR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EGR1 (early growth response 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen