Anti-HOXB9 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP36893_P050
AVIVARP36893_P050EA 401 EUR
AVIVARP36893_P050
Anti-HOXB9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HOXB9 (homeobox B9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen