Anti-CBFA2T2H Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP37525_P050
AVIVARP37525_P050EA 401 EUR
AVIVARP37525_P050
Anti-CBFA2T2H Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CBFA2T2H
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR