Anti-A830053O21Rik Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP37562_P050
AVIVARP37562_P050EA 401 EUR
AVIVARP37562_P050
Anti-A830053O21Rik Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: A830053O21Rik
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse