Anti-KLF11 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP37579_P050
AVIVARP37579_P050EA 401 EUR
AVIVARP37579_P050
Anti-KLF11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLF11 (Kruppel-like factor 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat