Anti-ETV7 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP37765_P050
AVIVARP37765_P050EA 401 EUR
AVIVARP37765_P050
Anti-ETV7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ETV7 (ets variant 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen