Anti-SCAND1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP37772_P050
AVIVARP37772_P050EA 401 EUR
AVIVARP37772_P050
Anti-SCAND1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SCAND1 (SCAN domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen