Anti-HMGN5 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP37788_P050
AVIVARP37788_P050EA 401 EUR
AVIVARP37788_P050
Anti-HMGN5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HMGN5 (high mobility group nucleosome binding domain 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR