Anti-NKX1-1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP37954_P050
AVIVARP37954_P050EA 401 EUR
AVIVARP37954_P050
Anti-NKX1-1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NKX1-1 (NK1 homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR