Anti-LMO3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP38139_P050
AVIVARP38139_P050EA 401 EUR
AVIVARP38139_P050
Anti-LMO3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LMO3 (LIM domain only 3 (rhombotin-like 2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen