Anti-SATB1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP38208_T100
AVIVARP38208_T100EA 349 EUR
AVIVARP38208_T100
Anti-SATB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SATB1 (SATB homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR