Anti-HLTF Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP38217_P050
AVIVARP38217_P050EA 401 EUR
AVIVARP38217_P050
Anti-HLTF Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HLTF (helicase-like transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen