Anti-TEF Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP38279_P050
AVIVARP38279_P050EA 401 EUR
AVIVARP38279_P050
Anti-TEF Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TEF (thyrotrophic embryonic factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen