Anti-BUD31 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP38402_T100
AVIVARP38402_T100EA 349 EUR
AVIVARP38402_T100
Anti-BUD31 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BUD31 (BUD31 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen