Anti-NFIC Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP38635_P050
AVIVARP38635_P050EA 401 EUR
AVIVARP38635_P050
Anti-NFIC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NFIC (nuclear factor I/C (CCAAT-binding transcription factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen