Anti-SOX1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP38720_P050
AVIVARP38720_P050EA 401 EUR
AVIVARP38720_P050
Anti-SOX1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SOX1 (SRY (sex determining region Y)-box 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen