Anti-TRIM16 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP38799_T100
AVIVARP38799_T100EA 349 EUR
AVIVARP38799_T100
Anti-TRIM16 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRIM16 (tripartite motif containing 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human