Anti-ZNF266 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP38843_P050
AVIVARP38843_P050EA 401 EUR
AVIVARP38843_P050
Anti-ZNF266 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF266 (zinc finger protein 266)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse