Anti-HOXD4 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP39116_T100
AVIVARP39116_T100EA 349 EUR
AVIVARP39116_T100
Anti-HOXD4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HOXD4 (homeobox D4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen