Anti-GTF2IRD1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP39253_T100
AVIVARP39253_T100EA 349 EUR
AVIVARP39253_T100
Anti-GTF2IRD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GTF2IRD1 (GTF2I repeat domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen