Anti-SIX4 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP39278_P050
AVIVARP39278_P050EA 401 EUR
AVIVARP39278_P050
Anti-SIX4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SIX4 (SIX homeobox 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen