Anti-ZNF529 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP39462_P050
AVIVARP39462_P050EA 401 EUR
AVIVARP39462_P050
Anti-ZNF529 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF529 (zinc finger protein 529)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Pig