Anti-FOXM1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP39518_P050
AVIVARP39518_P050EA 401 EUR
AVIVARP39518_P050
Anti-FOXM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FOXM1 (forkhead box M1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Dog, Human, Mouse, Pig, Rat