Anti-BTBD6 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP39756_T100
AVIVARP39756_T100EA 349 EUR
AVIVARP39756_T100
Anti-BTBD6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BTBD6 (BTB (POZ) domain containing 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen