Anti-ZNF709 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP39911_T100
AVIVARP39911_T100EA 349 EUR
AVIVARP39911_T100
Anti-ZNF709 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF709 (zinc finger protein 709)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR