Anti-POU5F2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP39945_T100
AVIVARP39945_T100EA 349 EUR
AVIVARP39945_T100
Anti-POU5F2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: POU5F2 (POU domain class 5, transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR