Anti-ZNF398 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP39969_P050
AVIVARP39969_P050EA 401 EUR
AVIVARP39969_P050
Anti-ZNF398 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF398 (zinc finger protein 398)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen