Anti-TCFAP2C Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP40141_T100
AVIVARP40141_T100EA 349 EUR
AVIVARP40141_T100
Anti-TCFAP2C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TCFAP2C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR