Anti-INSIG1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP40161_P050
AVIVARP40161_P050EA 401 EUR
AVIVARP40161_P050
Anti-INSIG1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: INSIG1 (insulin induced gene 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human