Anti-U2AF2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP40285_P050
AVIVARP40285_P050EA 401 EUR
AVIVARP40285_P050
Anti-U2AF2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: U2AF2 (U2 small nuclear RNA auxiliary factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen