Anti-HNRNPC Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP40517_P050
AVIVARP40517_P050EA 401 EUR
AVIVARP40517_P050
Anti-HNRNPC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HNRNPC (heterogeneous nuclear ribonucleoprotein C (C1/C2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen