Anti-RBM12 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP40622_P050
AVIVARP40622_P050EA 401 EUR
AVIVARP40622_P050
Anti-RBM12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RBM12 (RNA binding motif protein 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen