Anti-NXF1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP40638_P050
AVIVARP40638_P050EA 401 EUR
AVIVARP40638_P050
Anti-NXF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NXF1 (nuclear RNA export factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen