Anti-EXOSC6 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP41086_P050
AVIVARP41086_P050EA 401 EUR
AVIVARP41086_P050
Anti-EXOSC6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EXOSC6 (exosome component 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR