Anti-MAT1A Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP41399_T100
AVIVARP41399_T100EA 349 EUR
AVIVARP41399_T100
Anti-MAT1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAT1A (methionine adenosyltransferase I, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen